missing translation for 'onlineSavingsMsg'
Learn More

3-Bromomethyl-7-methoxy-1,4-benzoxazin-2-one 98.0+%, TCI America™

Catalog No. p-7118285
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A5553100MG 100 mg
A55531G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A5553100MG Supplier TCI America Supplier No. A5553100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 124522-09-4
Molecular Formula C10H8BrNO3
Molecular Weight (g/mol) 270.08
MDL Number MFCD00143099
InChI Key XFCZURAACWKKIH-UHFFFAOYSA-N
PubChem CID 195402
IUPAC Name 3-(bromomethyl)-7-methoxy-2H-1,4-benzoxazin-2-one
SMILES COC1=CC=C2N=C(CBr)C(=O)OC2=C1

Specifications

Quantity 100 mg
Formula Weight 270.08
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Bromomethyl-7-methoxy-1,4-benzoxazin-2-one [for HPLC Labeling]
TSCA No
Recommended Storage Refrigerator