accessibility menu, dialog, popup

UserName

CH3ONBrOO

CAS RN 124522-09-4

IUPAC Name: 3-(bromomethyl)-7-methoxy-2H-1,4-benzoxazin-2-one
Synonyms: 3-(bromomethyl)-7-methoxy-1,4-benzoxazin-2-one
Molecular Weight (g/mol): 270.08
Molecular Formula: C10H8BrNO3
InChi Key: XFCZURAACWKKIH-UHFFFAOYSA-N
SMILES: COC1=CC=C2N=C(CBr)C(=O)OC2=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results