missing translation for 'onlineSavingsMsg'
Learn More

3-(Bromomethyl)-5-chlorobenzo[b]thiophene 98.0+%, TCI America™

Catalog No. B31475G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B31475G 5 g
B31471G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B31475G Supplier TCI America Supplier No. B31475G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1198-51-2
Molecular Formula C9H6BrClS
Molecular Weight (g/mol) 261.56
MDL Number MFCD00173769
InChI Key FKQSFVITUNJLCY-UHFFFAOYSA-N
Synonym 3-bromomethyl-5-chlorobenzo b thiophene, 3-bromomethyl-5-chloro-1-benzothiophene, 3-bromomethyl-5-chloro benzo b thiophene, 3-bromomethyl-5-chlorobenzothiophene, 5-chloro-3-bromomethylbenzothiophene, 5-chloro-3-bromomethyl-benzo b-thiophene, benzo b thiophene, 3-bromomethyl-5-chloro, pubchem13553
PubChem CID 2779857
IUPAC Name 3-(bromomethyl)-5-chloro-1-benzothiophene
SMILES ClC1=CC=C2SC=C(CBr)C2=C1

Specifications

Melting Point 131°C
Quantity 5 g
UN Number 3261
Formula Weight 261.56
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-(Bromomethyl)-5-chlorobenzo[b]thiophene
TSCA No
Recommended Storage Refrigerator