missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Bromocatechol 98.0+%, TCI America™
Supplier: TCI America B37105G
Specifications
| 3-Bromocatechol | |
| 43°C | |
| 120°C | |
| MFCD00869768 | |
| 3-bromocatechol, 3-bromopyrocatechol, 1-bromo-2,3-dihydroxybenzene, 1,2-benzenediol, 3-bromo, benzenediol, bromo, 3-bromo-1,2-benzenediol, 2,3-dihydroxy-bromobenzene, bromocatechol, 3-bromoatechol, pubchem19085 | |
| C1=CC(=C(C(=C1)Br)O)O | |
| 189.008 | |
| 189.01 | |
| Crystalline Powder |
| 14381-51-2 | |
| Yellow | |
| C6H5BrO2 | |
| 5 g | |
| JPBDMIWPTFDFEU-UHFFFAOYSA-N | |
| 3-bromobenzene-1,2-diol | |
| 26659 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 14381-51-2 | |
| 189.008 | |
| JPBDMIWPTFDFEU-UHFFFAOYSA-N | |
| 26659 | |
| C1=CC(=C(C(=C1)Br)O)O |
| C6H5BrO2 | |
| MFCD00869768 | |
| 3-bromocatechol, 3-bromopyrocatechol, 1-bromo-2,3-dihydroxybenzene, 1,2-benzenediol, 3-bromo, benzenediol, bromo, 3-bromo-1,2-benzenediol, 2,3-dihydroxy-bromobenzene, bromocatechol, 3-bromoatechol, pubchem19085 | |
| 3-bromobenzene-1,2-diol |
Safety and Handling
TSCA : No