missing translation for 'onlineSavingsMsg'
Learn More

3-Bromocatechol 98.0+%, TCI America™

Catalog No. B37101G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B37101G 1 g
B37105G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B37101G Supplier TCI America Supplier No. B37101G

Chemical Identifiers

CAS 14381-51-2
Molecular Formula C6H5BrO2
Molecular Weight (g/mol) 189.008
MDL Number MFCD00869768
InChI Key JPBDMIWPTFDFEU-UHFFFAOYSA-N
Synonym 3-bromocatechol, 3-bromopyrocatechol, 1-bromo-2,3-dihydroxybenzene, 1,2-benzenediol, 3-bromo, benzenediol, bromo, 3-bromo-1,2-benzenediol, 2,3-dihydroxy-bromobenzene, bromocatechol, 3-bromoatechol, pubchem19085
PubChem CID 26659
IUPAC Name 3-bromobenzene-1,2-diol
SMILES C1=CC(=C(C(=C1)Br)O)O

Specifications

Melting Point 43°C
Color Yellow
Boiling Point 120°C
Quantity 1 g
Formula Weight 189.01
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Bromocatechol