missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Bromocatechol, 95%
Description
3-Bromocatechol, is used as a drug and agrochemicals intermediate, Pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Bromocatechol, is used as a drug and agrochemicals intermediate, Pharmaceutical intermediate.
Solubility
Soluble in organic solvents.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents.
3-Bromocatechol, is used as a drug and agrochemicals intermediate, Pharmaceutical intermediate.
Solubility
Soluble in organic solvents.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents.
Specifications
Specifications
| Melting Point | 39°C to 42°C |
| Quantity | 250 mg |
| Solubility Information | Soluble in organic solvents. |
| Formula Weight | 189.01 |
| Percent Purity | 95% |
| Chemical Name or Material | 3-Bromocatechol |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only