missing translation for 'onlineSavingsMsg'
Learn More

3-Bromobenzylamine 98.0+%, TCI America™

Catalog No. p-7123685
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B46765G 5 g
B467625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B46765G Supplier TCI America Supplier No. B46765G
Only null left

Chemical Identifiers

CAS 10269-01-9
Molecular Formula C7H8BrN
Molecular Weight (g/mol) 186.052
MDL Number MFCD01026119
InChI Key SUYJXERPRICYRX-UHFFFAOYSA-N
Synonym 3-bromobenzylamine, 3-bromophenyl methanamine, benzenemethanamine, 3-bromo, 3-bromophenyl methylamine, m-bromobenzylamine, 1-3-bromophenyl methanamine, 3-bromo-benzylamine, 3-bromobenzenemethanamine, 3-bromobenzyl amine, pubchem21109
PubChem CID 457587
IUPAC Name (3-bromophenyl)methanamine
SMILES C1=CC(=CC(=C1)Br)CN

Specifications

Color Yellow
Boiling Point 245°C
Quantity 5 g
UN Number 2735
Formula Weight 186.05
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material 3-Bromobenzylamine