accessibility menu, dialog, popup

UserName

CAS RN 10269-01-9

IUPAC Name: 1-(3-bromophenyl)methanamine
Synonyms: 1-(3-bromophenyl)methanamine
Molecular Weight (g/mol): 186.05
Molecular Formula: C7H8BrN
InChi Key: SUYJXERPRICYRX-UHFFFAOYSA-N
SMILES: NCC1=CC=CC(Br)=C1
Molecular Weight (g/mol) 
  • (3)
  • (5)
Percent Purity 
  • (2)
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (6)
  • (3)
  • (5)
  • (2)
  • (3)
  • (3)
  • (1)
  • (1)
  • (3)
  • (3)
  • (5)
  • (2)