missing translation for 'onlineSavingsMsg'
Learn More

3-Bromobenzylamine Hydrochloride 97.0+%, TCI America™

Catalog No. b19895g
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
5 g
Catalog No. Quantity
B19895G 5 g
B198925G 25 g
2 options

Catalog No. B19895G

Supplier: TCI America B19895G

Only null left

Chemical Identifiers

CAS 39959-54-1
Molecular Formula C7H9BrClN
Molecular Weight (g/mol) 222.51
MDL Number MFCD00012856
InChI Key UGEQUCUBWNAUJS-UHFFFAOYSA-N
Synonym 3-bromobenzylamine hydrochloride, 3-bromophenyl methanamine hydrochloride, 3-bromobenzylamine hcl, 3-bromobenzylaminehydrochloride, m-bromobenzylamine hydrochloride, 3-bromophenyl methylamine hydrochloride, 1-3-bromophenyl methanamine hydrochloride, benzenemethanamine, 3-bromo-, hydrochloride, pubchem3750, 3-bromobenzyl amine hcl
PubChem CID 2724980
IUPAC Name (3-bromophenyl)methanamine;hydrochloride
SMILES C1=CC(=CC(=C1)Br)CN.Cl

Specifications

Melting Point 220°C
Color White-Yellow
Quantity 5 g
Formula Weight 222.51
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Bromobenzylamine Hydrochloride