accessibility menu, dialog, popup

UserName

CAS RN 39959-54-1

IUPAC Name: (3-bromophenyl)methanaminium
Synonyms: (3-bromophenyl)methanaminium
Molecular Weight (g/mol): 187.06
Molecular Formula: C7H9BrN
InChi Key: SUYJXERPRICYRX-UHFFFAOYSA-O
SMILES: [NH3+]CC1=CC=CC(Br)=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)