missing translation for 'onlineSavingsMsg'
Learn More

3-Bromobenzylamine hydrochloride, 98%

Catalog No. AAA1613214
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1613214 25 g
AAA1613206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1613214 Supplier Thermo Scientific Chemicals Supplier No. A1613214
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 39959-54-1 | C7H9BrClN | 222.51 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 39959-54-1
Molecular Formula C7H9BrClN
Molecular Weight (g/mol) 222.51
MDL Number MFCD00012856
InChI Key UGEQUCUBWNAUJS-UHFFFAOYSA-N
Synonym 3-bromobenzylamine hydrochloride, 3-bromophenyl methanamine hydrochloride, 3-bromobenzylamine hcl, 3-bromobenzylaminehydrochloride, m-bromobenzylamine hydrochloride, 3-bromophenyl methylamine hydrochloride, 1-3-bromophenyl methanamine hydrochloride, benzenemethanamine, 3-bromo-, hydrochloride, pubchem3750, 3-bromobenzyl amine hcl
PubChem CID 2724980
IUPAC Name (3-bromophenyl)methanamine;hydrochloride
SMILES C1=CC(=CC(=C1)Br)CN.Cl

Specifications

Melting Point 218°C to 221°C
Quantity 25 g
Beilstein 3913185
Sensitivity Hygroscopic
Formula Weight 222.52
Percent Purity 98%
Chemical Name or Material 3-Bromobenzylamine hydrochloride
Hazard Category H302+H312-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only