missing translation for 'onlineSavingsMsg'
Learn More

3-Bromobenzyl Chloride 98.0+%, TCI America™

Catalog No. p-7119556
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B22135G 5 g
B221325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B22135G Supplier TCI America Supplier No. B22135G

Chemical Identifiers

CAS 932-77-4
Molecular Formula C7H6BrCl
Molecular Weight (g/mol) 205.479
MDL Number MFCD00040865
InChI Key UDKGXKYEWBGQCG-UHFFFAOYSA-N
Synonym 3-bromobenzyl chloride, 1-bromo-3-chloromethyl benzene, 3-bromo-alpha-chlorotoluene, m-bromobenzyl chloride, benzene, 1-bromo-3-chloromethyl, 3-bromo-a-chlorotoluene, 3-bromobenzylchloride, pubchem23319, acmc-209rkw, bromo-3-chloromethyl-benzene
PubChem CID 523059
IUPAC Name 1-bromo-3-(chloromethyl)benzene
SMILES C1=CC(=CC(=C1)Br)CCl

Specifications

Color Yellow
Quantity 5 g
UN Number 3265
Formula Weight 205.48
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Bromobenzyl Chloride