missing translation for 'onlineSavingsMsg'
Learn More

3-Bromobenzyl chloride, 97%

Catalog No. AAL1245806
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1245806 5 g
AAL1245814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1245806 Supplier Thermo Scientific Chemicals Supplier No. L1245806
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 932-77-4 | C7H6BrCl | 205.479 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 932-77-4
Molecular Formula C7H6BrCl
Molecular Weight (g/mol) 205.479
MDL Number MFCD00040865
InChI Key UDKGXKYEWBGQCG-UHFFFAOYSA-N
Synonym 3-bromobenzyl chloride, 1-bromo-3-chloromethyl benzene, 3-bromo-alpha-chlorotoluene, m-bromobenzyl chloride, benzene, 1-bromo-3-chloromethyl, 3-bromo-a-chlorotoluene, 3-bromobenzylchloride, pubchem23319, acmc-209rkw, bromo-3-chloromethyl-benzene
PubChem CID 523059
IUPAC Name 1-bromo-3-(chloromethyl)benzene
SMILES C1=CC(=CC(=C1)Br)CCl

Specifications

Density 1.608
Boiling Point 118°C to 119°C (18 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.5835
Quantity 5 g
UN Number UN3265
Beilstein 742267
Formula Weight 205.49
Percent Purity 97%
Chemical Name or Material 3-Bromobenzyl chloride
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only