Learn More
3-Bromobenzyl bromide, 99%
CAS: 823-78-9 | C7H6Br2 | 249.933 g/mol
Supplier: Thermo Scientific Chemicals A1507518
Description
3-Bromobenzyl bromide was used in the synthesis of, 1,7-di(3-bromobenzyl)cyclen and substituted 8-arylquinoline, phosphodiesterase 4 (PDE4) inhibitors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-Bromobenzyl bromide was used in the synthesis of, 1,7-di(3-bromobenzyl)cyclen and substituted 8-arylquinoline, phosphodiesterase 4 (PDE4) inhibitors.
Solubility
Insoluble in water.
Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
Specifications
| 3-Bromobenzyl bromide | |
| 40°C to 44°C | |
| 128°C to 130°C (12 mmHg) | |
| C7H6Br2 | |
| 50 g | |
| 2078683 | |
| Insoluble in water. | |
| C1=CC(=CC(=C1)Br)CBr | |
| 249.933 | |
| 249.94 |
| 823-78-9 | |
| 1.56 | |
| >110°C (230°F) | |
| MFCD00000176 | |
| UN1759 | |
| 3-bromobenzyl bromide, 1-bromo-3-bromomethyl benzene, m-bromobenzyl bromide, 3-bromobenzylbromide, benzene, 1-bromo-3-bromomethyl, alpha,3-dibromotoluene, alpha,m-dibromotoluene, 3-bromo benzyl bromide, 1-bromo-3-bromomethyl-benzene, alpha-3-dibromotoluene | |
| ZPCJPJQUVRIILS-UHFFFAOYSA-N | |
| 1-bromo-3-(bromomethyl)benzene | |
| 69979 | |
| 99% |
Chemical Identifiers
| 823-78-9 | |
| 249.933 | |
| ZPCJPJQUVRIILS-UHFFFAOYSA-N | |
| 69979 | |
| C1=CC(=CC(=C1)Br)CBr |
| C7H6Br2 | |
| MFCD00000176 | |
| 3-bromobenzyl bromide, 1-bromo-3-bromomethyl benzene, m-bromobenzyl bromide, 3-bromobenzylbromide, benzene, 1-bromo-3-bromomethyl, alpha,3-dibromotoluene, alpha,m-dibromotoluene, 3-bromo benzyl bromide, 1-bromo-3-bromomethyl-benzene, alpha-3-dibromotoluene | |
| 1-bromo-3-(bromomethyl)benzene |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLIDS, N.O.S.
EINECSNumber : 212-519-1
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only