missing translation for 'onlineSavingsMsg'
Learn More

3-Bromobenzyl bromide, 99%

Catalog No. p-7022350
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1507509 10 g
AAA1507518 50 g
AAA1507530 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1507509 Supplier Thermo Scientific Chemicals Supplier No. A1507509

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 823-78-9 | C7H6Br2 | 249.933 g/mol

3-Bromobenzyl bromide was used in the synthesis of, 1,7-di(3-bromobenzyl)cyclen and substituted 8-arylquinoline, phosphodiesterase 4 (PDE4) inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Bromobenzyl bromide was used in the synthesis of, 1,7-di(3-bromobenzyl)cyclen and substituted 8-arylquinoline, phosphodiesterase 4 (PDE4) inhibitors.

Solubility
Insoluble in water.

Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.

Chemical Identifiers

CAS 823-78-9
Molecular Formula C7H6Br2
Molecular Weight (g/mol) 249.933
MDL Number MFCD00000176
InChI Key ZPCJPJQUVRIILS-UHFFFAOYSA-N
Synonym 3-bromobenzyl bromide, 1-bromo-3-bromomethyl benzene, m-bromobenzyl bromide, 3-bromobenzylbromide, benzene, 1-bromo-3-bromomethyl, alpha,3-dibromotoluene, alpha,m-dibromotoluene, 3-bromo benzyl bromide, 1-bromo-3-bromomethyl-benzene, alpha-3-dibromotoluene
PubChem CID 69979
IUPAC Name 1-bromo-3-(bromomethyl)benzene
SMILES C1=CC(=CC(=C1)Br)CBr

Specifications

Melting Point 40°C to 44°C
Density 1.56
Boiling Point 128°C to 130°C (12 mmHg)
Flash Point >110°C (230°F)
Quantity 10 g
UN Number UN1759
Beilstein 2078683
Solubility Information Insoluble in water.
Formula Weight 249.94
Percent Purity 99%
Chemical Name or Material 3-Bromobenzyl bromide
Show More Show Less
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLIDS, N.O.S.
EINECSNumber 212-519-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only