missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Bromobenzotrifluoride 99.0+%, TCI America™
$29.83 - $190.13
Chemical Identifiers
| CAS | 401-78-5 |
|---|---|
| Molecular Formula | C7H4BrF3 |
| Molecular Weight (g/mol) | 225.008 |
| MDL Number | MFCD00000380 |
| InChI Key | NNMBNYHMJRJUBC-UHFFFAOYSA-N |
| Synonym | 3-bromobenzotrifluoride, 1-bromo-3-trifluoromethyl benzene, m-bromobenzotrifluoride, 3-trifluoromethyl bromobenzene, benzene, 1-bromo-3-trifluoromethyl, 3-brombenzotrifluorid, 3-bromotrifluoromethylbenzene, 3-bromobenzyltrifluoride, 3-trifluoromethyl phenyl bromide, m-trifluoromethyl bromobenzene |
| PubChem CID | 9817 |
| IUPAC Name | 1-bromo-3-(trifluoromethyl)benzene |
| SMILES | C1=CC(=CC(=C1)Br)C(F)(F)F |
Chemical Identifiers
| 401-78-5 | |
| 225.008 | |
| NNMBNYHMJRJUBC-UHFFFAOYSA-N | |
| 9817 | |
| C1=CC(=CC(=C1)Br)C(F)(F)F |
| C7H4BrF3 | |
| MFCD00000380 | |
| 3-bromobenzotrifluoride, 1-bromo-3-trifluoromethyl benzene, m-bromobenzotrifluoride, 3-trifluoromethyl bromobenzene, benzene, 1-bromo-3-trifluoromethyl, 3-brombenzotrifluorid, 3-bromotrifluoromethylbenzene, 3-bromobenzyltrifluoride, 3-trifluoromethyl phenyl bromide, m-trifluoromethyl bromobenzene | |
| 1-bromo-3-(trifluoromethyl)benzene |
Specifications
| 401-78-5 | |
| C7H4BrF3 | |
| 25 g | |
| 3-bromobenzotrifluoride, 1-bromo-3-trifluoromethyl benzene, m-bromobenzotrifluoride, 3-trifluoromethyl bromobenzene, benzene, 1-bromo-3-trifluoromethyl, 3-brombenzotrifluorid, 3-bromotrifluoromethylbenzene, 3-bromobenzyltrifluoride, 3-trifluoromethyl phenyl bromide, m-trifluoromethyl bromobenzene | |
| C1=CC(=CC(=C1)Br)C(F)(F)F | |
| 225.008 | |
| 225.01 | |
| Liquid |
| 155°C | |
| MFCD00000380 | |
| 1993 | |
| NNMBNYHMJRJUBC-UHFFFAOYSA-N | |
| 1-bromo-3-(trifluoromethyl)benzene | |
| 9817 | |
| ≥99.0% (GC) | |
| 3-Bromobenzotrifluoride |
Safety and Handling
EINECSNumber : (3)-0068&(9)-1242
RTECSNumber : XS7970000
TSCA : Yes