missing translation for 'onlineSavingsMsg'
Learn More

3-Bromobenzotrifluoride, 98+%

Catalog No. AAA1472414
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1472414 25 g
AAA1472422 100 g
AAA1472436 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1472414 Supplier Thermo Scientific Chemicals Supplier No. A1472414
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 401-78-5 | C7H4BrF3 | 225.008 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 401-78-5
Molecular Formula C7H4BrF3
Molecular Weight (g/mol) 225.008
MDL Number MFCD00000380
InChI Key NNMBNYHMJRJUBC-UHFFFAOYSA-N
Synonym 3-bromobenzotrifluoride, 1-bromo-3-trifluoromethyl benzene, m-bromobenzotrifluoride, 3-trifluoromethyl bromobenzene, benzene, 1-bromo-3-trifluoromethyl, 3-brombenzotrifluorid, 3-bromotrifluoromethylbenzene, 3-bromobenzyltrifluoride, 3-trifluoromethyl phenyl bromide, m-trifluoromethyl bromobenzene
PubChem CID 9817
IUPAC Name 1-bromo-3-(trifluoromethyl)benzene
SMILES C1=CC(=CC(=C1)Br)C(F)(F)F

Specifications

Density 1.613
Boiling Point 151°C to 152°C
Flash Point 59°C (138°F)
Odor Aromatic
Refractive Index 1.473
Quantity 25 g
UN Number UN1993
Beilstein 1449557
Formula Weight 225.01
Percent Purity ≥98%
Chemical Name or Material 3-Bromobenzotrifluoride
Show More Show Less
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 206-932-6
RTECSNumber XS7970000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only