missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Bromobenzonitrile 98.0+%, TCI America™
$63.52 - $446.64
Chemical Identifiers
| CAS | 6952-59-6 |
|---|---|
| Molecular Formula | C7H4BrN |
| Molecular Weight (g/mol) | 182.02 |
| MDL Number | MFCD00001796 |
| InChI Key | STXAVEHFKAXGOX-UHFFFAOYSA-N |
| Synonym | m-bromobenzonitrile, benzonitrile, 3-bromo, benzonitrile, m-bromo, 3-cyanobromobenzene, 1-bromo-3-cyanobenzene, 3-bromobenzenecarbonitrile, 3-bromo benzonitrile, 3-bromobenzonitride, m-brombenzonitril, 3-brombenzonitrile |
| PubChem CID | 23381 |
| IUPAC Name | 3-bromobenzonitrile |
| SMILES | BrC1=CC=CC(=C1)C#N |
Chemical Identifiers
| 6952-59-6 | |
| 182.02 | |
| STXAVEHFKAXGOX-UHFFFAOYSA-N | |
| 23381 | |
| BrC1=CC=CC(=C1)C#N |
| C7H4BrN | |
| MFCD00001796 | |
| m-bromobenzonitrile, benzonitrile, 3-bromo, benzonitrile, m-bromo, 3-cyanobromobenzene, 1-bromo-3-cyanobenzene, 3-bromobenzenecarbonitrile, 3-bromo benzonitrile, 3-bromobenzonitride, m-brombenzonitril, 3-brombenzonitrile | |
| 3-bromobenzonitrile |
Specifications
| 6952-59-6 | |
| White-Yellow | |
| C7H4BrN | |
| 5 g | |
| m-bromobenzonitrile, benzonitrile, 3-bromo, benzonitrile, m-bromo, 3-cyanobromobenzene, 1-bromo-3-cyanobenzene, 3-bromobenzenecarbonitrile, 3-bromo benzonitrile, 3-bromobenzonitride, m-brombenzonitril, 3-brombenzonitrile | |
| BrC1=CC=CC(=C1)C#N | |
| 182.02 | |
| 182.02 | |
| Crystalline Powder |
| 40°C | |
| 225°C | |
| MFCD00001796 | |
| 3439 | |
| STXAVEHFKAXGOX-UHFFFAOYSA-N | |
| 3-bromobenzonitrile | |
| 23381 | |
| ≥98.0% (GC) | |
| 3-Bromobenzonitrile |
Safety and Handling
RTECSNumber : DI2459500
TSCA : No