missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Bromobenzoic Acid 98.0+%, TCI America™
$106.25 - $590.90
Chemical Identifiers
| CAS | 585-76-2 |
|---|---|
| Molecular Formula | C7H5BrO2 |
| Molecular Weight (g/mol) | 201.019 |
| MDL Number | MFCD00002487 |
| InChI Key | VOIZNVUXCQLQHS-UHFFFAOYSA-N |
| Synonym | m-bromobenzoic acid, 3-brombenzoic acid, benzoic acid, 3-bromo, 3-bromo-benzoic acid, benzoic acid, m-bromo, unii-7dct3a73lx, 7dct3a73lx, 3-bromo benzene acid, 3-bromobenzoicacid, 4-fluorobenzoicacid |
| PubChem CID | 11456 |
| IUPAC Name | 3-bromobenzoic acid |
| SMILES | C1=CC(=CC(=C1)Br)C(=O)O |
Chemical Identifiers
| 585-76-2 | |
| 201.019 | |
| VOIZNVUXCQLQHS-UHFFFAOYSA-N | |
| 11456 | |
| C1=CC(=CC(=C1)Br)C(=O)O |
| C7H5BrO2 | |
| MFCD00002487 | |
| m-bromobenzoic acid, 3-brombenzoic acid, benzoic acid, 3-bromo, 3-bromo-benzoic acid, benzoic acid, m-bromo, unii-7dct3a73lx, 7dct3a73lx, 3-bromo benzene acid, 3-bromobenzoicacid, 4-fluorobenzoicacid | |
| 3-bromobenzoic acid |
Specifications
| 585-76-2 | |
| White-Yellow | |
| MFCD00002487 | |
| m-bromobenzoic acid, 3-brombenzoic acid, benzoic acid, 3-bromo, 3-bromo-benzoic acid, benzoic acid, m-bromo, unii-7dct3a73lx, 7dct3a73lx, 3-bromo benzene acid, 3-bromobenzoicacid, 4-fluorobenzoicacid | |
| C1=CC(=CC(=C1)Br)C(=O)O | |
| 201.019 | |
| 201.02 | |
| Crystalline Powder |
| 158°C | |
| C7H5BrO2 | |
| 25 g | |
| VOIZNVUXCQLQHS-UHFFFAOYSA-N | |
| 3-bromobenzoic acid | |
| 11456 | |
| ≥98.0% (GC,T) | |
| 3-Bromobenzoic Acid |
Safety and Handling
EINECSNumber : (3)-0062
TSCA : Yes