Learn More
3-Bromobenzoic acid, 98+%
CAS: 585-76-2 | C7H5BrO2 | 201.019 g/mol
$113.86 - $400.73
Chemical Identifiers
| CAS | 585-76-2 |
|---|---|
| Molecular Formula | C7H5BrO2 |
| Molecular Weight (g/mol) | 201.019 |
| MDL Number | MFCD00002487 |
| InChI Key | VOIZNVUXCQLQHS-UHFFFAOYSA-N |
| Synonym | m-bromobenzoic acid, 3-brombenzoic acid, benzoic acid, 3-bromo, 3-bromo-benzoic acid, benzoic acid, m-bromo, unii-7dct3a73lx, 7dct3a73lx, 3-bromo benzene acid, 3-bromobenzoicacid, 4-fluorobenzoicacid |
| PubChem CID | 11456 |
| IUPAC Name | 3-bromobenzoic acid |
| SMILES | C1=CC(=CC(=C1)Br)C(=O)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1260614
|
Thermo Scientific Chemicals
A1260614 |
25 g |
Each for $113.86
|
|
|||||
|
AAA1260622
|
Thermo Scientific Chemicals
A1260622 |
100 g |
Each for $400.73
|
|
|||||
Description
3-Bromobenzoic acid was used as test solute in the determination of acidity constants by capillary zone electrophoresis. It was used as internal standard to study the retention mechanisms of an unmodified and a hydroxylated polystyrene-divinylbenzene polymer by solid-phase extraction. It was also used in synthesis of N-(1,1-dimethyl-2-hydroxyethyl)-3-bromobenzamide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-Bromobenzoic acid was used as test solute in the determination of acidity constants by capillary zone electrophoresis. It was used as internal standard to study the retention mechanisms of an unmodified and a hydroxylated polystyrene-divinylbenzene polymer by solid-phase extraction. It was also used in synthesis of N-(1,1-dimethyl-2-hydroxyethyl)-3-bromobenzamide.
Solubility
Insoluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Chemical Identifiers
| 585-76-2 | |
| 201.019 | |
| VOIZNVUXCQLQHS-UHFFFAOYSA-N | |
| 11456 | |
| C1=CC(=CC(=C1)Br)C(=O)O |
| C7H5BrO2 | |
| MFCD00002487 | |
| m-bromobenzoic acid, 3-brombenzoic acid, benzoic acid, 3-bromo, 3-bromo-benzoic acid, benzoic acid, m-bromo, unii-7dct3a73lx, 7dct3a73lx, 3-bromo benzene acid, 3-bromobenzoicacid, 4-fluorobenzoicacid | |
| 3-bromobenzoic acid |
Specifications
| 585-76-2 | |
| 1.948 | |
| MFCD00002487 | |
| 2041792 | |
| Insoluble in water. | |
| C1=CC(=CC(=C1)Br)C(=O)O | |
| 201.019 | |
| 201.02 | |
| 3-Bromobenzoic acid |
| 153°C to 157°C | |
| C7H5BrO2 | |
| 25 g | |
| m-bromobenzoic acid, 3-brombenzoic acid, benzoic acid, 3-bromo, 3-bromo-benzoic acid, benzoic acid, m-bromo, unii-7dct3a73lx, 7dct3a73lx, 3-bromo benzene acid, 3-bromobenzoicacid, 4-fluorobenzoicacid | |
| VOIZNVUXCQLQHS-UHFFFAOYSA-N | |
| 3-bromobenzoic acid | |
| 11456 | |
| ≥98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 209-562-3
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only