missing translation for 'onlineSavingsMsg'
Learn More

3-Bromobenzaldehyde 98.0+%, TCI America™

Catalog No. p-7118515
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Catalog No. Quantity
B054825G 25 g
B0548100G 100 g
B0548500G 500 g
3 options

Catalog No. B054825G

Supplier: TCI America B054825G

Only null left

Chemical Identifiers

CAS 3132-99-8
Molecular Formula C7H5BrO
Molecular Weight (g/mol) 185.02
MDL Number MFCD00003345
InChI Key SUISZCALMBHJQX-UHFFFAOYSA-N
Synonym m-bromobenzaldehyde, benzaldehyde, 3-bromo, benzaldehyde, m-bromo, 3-bromobenzeldehyde, unii-w0ard2zj82, 3-bromo benzaldehyde, 3-bromo-benzaldehyde, w0ard2zj82, benzaldehyde, m-bromo-6ci,7ci,8ci, 3-bromobenzaidehyde
PubChem CID 76583
IUPAC Name 3-bromobenzaldehyde
SMILES C1=CC(=CC(=C1)Br)C=O

Specifications

Color Yellow
Boiling Point 236°C
Quantity 25 g
Formula Weight 185.02
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Bromobenzaldehyde
EINECSNumber (3)-4111
RTECSNumber CU4820000
TSCA Yes