accessibility menu, dialog, popup

UserName

CAS RN 3132-99-8

IUPAC Name: 3-bromobenzaldehyde
Synonyms: 3-bromobenzaldehyde
Molecular Weight (g/mol): 185.02
Molecular Formula: C7H5BrO
InChi Key: SUISZCALMBHJQX-UHFFFAOYSA-N
SMILES: BrC1=CC=CC(C=O)=C1
Percent Purity 
  • (3)
  • (2)
  • (2)
  • (3)
Boiling Point 
  • (2)
  • (3)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
Physical Form 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (3)
  • (5)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (3)
  • (2)
  • (2)