missing translation for 'onlineSavingsMsg'
Learn More

3-Bromobenzaldehyde, 96%

Catalog No. AC106660250
Change view
Click to view available options
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC106660250 25 mL Glass bottle
AC106661000 100 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC106660250 Supplier Thermo Scientific Chemicals Supplier No. 106660250

CAS: 3132-99-8 | C7H5BrO | 185.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3132-99-8
Molecular Formula C7H5BrO
Molecular Weight (g/mol) 185.02
MDL Number MFCD00003345
InChI Key SUISZCALMBHJQX-UHFFFAOYSA-N
Synonym m-bromobenzaldehyde, benzaldehyde, 3-bromo, benzaldehyde, m-bromo, 3-bromobenzeldehyde, unii-w0ard2zj82, 3-bromo benzaldehyde, 3-bromo-benzaldehyde, w0ard2zj82, benzaldehyde, m-bromo-6ci,7ci,8ci, 3-bromobenzaidehyde
PubChem CID 76583
IUPAC Name 3-bromobenzaldehyde
SMILES C1=CC(=CC(=C1)Br)C=O

Specifications

CAS Min % 95.0
CAS Max % 100.0
Melting Point 18.0°C to 21.0°C
Color Colorless to Yellow
Density 1.5800g/mL
Boiling Point 228.0°C to 230.0°C
Flash Point 96°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Refractive Index 1.5920 to 1.5960
Linear Formula BrC6H4CHO
Quantity 25 mL
Beilstein 07, 238
Specific Gravity 1.58
Solubility Information Solubility in water: decomposes. Other solubilities: soluble in most organic solvents
Formula Weight 185.02
Percent Purity 96%
Physical Form Liquid After Melting
Chemical Name or Material 3-Bromobenzaldehyde, 96%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove
EINECSNumber 221-526-9
RTECSNumber CU4820000
TSCA TSCA

RUO – Research Use Only