missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-5'-phenyl-1,1':3',1″-terphenyl 98.0+%, TCI America™

Catalog No. p-7123818
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B48881G 1 g
B48885G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B48881G Supplier TCI America Supplier No. B48881G
Only null left

Chemical Identifiers

CAS 1233200-57-1
Molecular Formula C24H17Br
Molecular Weight (g/mol) 385.304
MDL Number MFCD28134530
InChI Key RDSDKECSKZPOLF-UHFFFAOYSA-N
PubChem CID 66797608
IUPAC Name 1-(3-bromophenyl)-3,5-diphenylbenzene
SMILES C1=CC=C(C=C1)C2=CC(=CC(=C2)C3=CC(=CC=C3)Br)C4=CC=CC=C4

Specifications

Melting Point 114°C
Color White-Yellow
Quantity 1 g
Synonym 5′C-(3-Bromophenyl)-1,1′C:3′C,1′C′C-terphenyl, 3-Bromo-5′C-phenyl-m-terphenyl, 5′C-(3-Bromophenyl)-m-terphenyl
Formula Weight 385.30
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Bromo-5′-phenyl-1,1′:3′,1′′-terphenyl