accessibility menu, dialog, popup

UserName

CAS RN 1233200-57-1

Br

CAS RN 1233200-57-1

IUPAC Name: 3-(3-bromophenyl)-5-phenyl-1,1'-biphenyl
Synonyms: 3-(3-bromophenyl)-5-phenyl-1,1'-biphenyl
Molecular Weight (g/mol): 385.3
Molecular Formula: C24H17Br
InChi Key: RDSDKECSKZPOLF-UHFFFAOYSA-N
SMILES: BrC1=CC=CC(=C1)C1=CC(=CC(=C1)C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results