missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Bromo-5'-phenyl-1,1':3',1″-terphenyl 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 114°C |
| Color | White-Yellow |
| Quantity | 1 g |
| Synonym | 5′C-(3-Bromophenyl)-1,1′C:3′C,1′C′C-terphenyl, 3-Bromo-5′C-phenyl-m-terphenyl, 5′C-(3-Bromophenyl)-m-terphenyl |
| Formula Weight | 385.30 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 3-Bromo-5′-phenyl-1,1′:3′,1′′-terphenyl |
Safety and Handling
| TSCA | No |