missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-5-nitroanisole 98.0+%, TCI America™

Catalog No. B49735G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B49735G 5 g
B49731G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B49735G Supplier TCI America Supplier No. B49735G
Only null left

Chemical Identifiers

CAS 16618-67-0
Molecular Formula C7H6BrNO3
Molecular Weight (g/mol) 232.033
MDL Number MFCD04112752
InChI Key MEQKSFQEPDRNEQ-UHFFFAOYSA-N
Synonym 3-bromo-5-nitroanisole, 3-bromo-5-nitroanisol, 5-bromo-1-methoxy-3-nitrobenzene, 1-bromo-3-methoxy-5-nitro-benzene, benzene, 1-bromo-3-methoxy-5-nitro, pubchem21624, acmc-209dts, 3-bromo-5-methoxynitrobenzene, benzene,1-bromo-3-methoxy-5-nitro, 3-methoxy-5-nitrophenyl bromide
PubChem CID 12299076
IUPAC Name 1-bromo-3-methoxy-5-nitrobenzene
SMILES COC1=CC(=CC(=C1)[N+](=O)[O-])Br

Specifications

Melting Point 88°C
Color Yellow
Quantity 5 g
Formula Weight 232.03
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Bromo-5-nitroanisole