accessibility menu, dialog, popup

UserName

CH3ONOOBr

CAS RN 16618-67-0

IUPAC Name: 1-bromo-3-methoxy-5-nitrobenzene
Synonyms: 1-bromo-3-methoxy-5-nitrobenzene
Molecular Weight (g/mol): 232.03
Molecular Formula: C7H6BrNO3
InChi Key: MEQKSFQEPDRNEQ-UHFFFAOYSA-N
SMILES: COC1=CC(Br)=CC(=C1)[N+]([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results