missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-5-iodobenzoic Acid 98.0+%, TCI America™

Catalog No. p-7119277
Change view
Click to view available options
Quantity:
5 g
25 g
Catalog No. Quantity
B18075G 5 g
B180725G 25 g
2 options

Catalog No. B18075G

Supplier: TCI America B18075G

Only null left

Chemical Identifiers

CAS 188815-32-9
Molecular Formula C7H4BrIO2
Molecular Weight (g/mol) 326.915
MDL Number MFCD00191851
InChI Key MKJBJYCBKXPQSY-UHFFFAOYSA-N
Synonym 5-bromo-3-iodobenzoic acid, 3-bromo-5-iodo-benzoic acid, 3-bromo-5-iodobenzoicacid, benzoic acid, 3-bromo-5-iodo, 3-iodo-5-bromobenzoic acid, pubchem3788, acmc-209erv, 3-bromo-5-iodobenzoic aicd, 3-bromo-5-iodo benzoic acid, 5-iodo-3-bromo-benzoic acid
PubChem CID 5182520
IUPAC Name 3-bromo-5-iodobenzoic acid
SMILES C1=C(C=C(C=C1Br)I)C(=O)O

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 326.92
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Bromo-5-iodobenzoic Acid