accessibility menu, dialog, popup

UserName

CAS RN 188815-32-9

IUPAC Name: 3-bromo-5-iodobenzoate
Synonyms: 3-bromo-5-iodobenzoate
Molecular Weight (g/mol): 325.91
Molecular Formula: C7H3BrIO2
InChi Key: MKJBJYCBKXPQSY-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1=CC(Br)=CC(I)=C1
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)