missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-5-iodobenzoic acid, 97%

Catalog No. AAA1707622
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1707622 100 g
AAA1707606 5 g
AAA1707614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1707622 Supplier Thermo Scientific Chemicals Supplier No. A1707622
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 188815-32-9 | C7H4BrIO2 | 326.915 g/mol

3-Bromo-5-iodobenzoic acid may be used in the preparation of the following:

  • Phenyl(3-bromo-5-iodo)benzoate.
  • As starting reagent for the large-scale synthesis of the thromboxane receptor antagonist 3-{3-[2-(4-chlorobenzenesulfonamido)ethyl]-5-(4-fluorobenzyl)phenyl}propionic acid, via regioselective Heck cross-coupling reaction.
  • Methyl 3-bromo-5-iodobenzoate.
  • 3-Bromo-5-(triisopropylsilylethynyl)benzoic acid, via Sonogashira coupling reaction.
  • Trifluoroacetophenone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Bromo-5-iodobenzoic acid may be used in the preparation of the following: • Phenyl(3-bromo-5-iodo)benzoate. • As starting reagent for the large-scale synthesis of the thromboxane receptor antagonist 3-{3-[2-(4-chlorobenzenesulfonamido)ethyl]-5-(4-fluorobenzyl)phenyl}propionic acid, via regioselective Heck cross-coupling reaction. • Methyl 3-bromo-5-iodobenzoate. • 3-Bromo-5-(triisopropylsilylethynyl)benzoic acid, via Sonogashira coupling reaction. • Trifluoroacetophenone.

Solubility
Soluble in methanol. Sparingly Soluble (0.20 g/L) (25°C), Calc.

Notes
Light Sensitive. Protect from light. Store away from oxidizing agents.

Chemical Identifiers

CAS 188815-32-9
Molecular Formula C7H4BrIO2
Molecular Weight (g/mol) 326.915
MDL Number MFCD00191851
InChI Key MKJBJYCBKXPQSY-UHFFFAOYSA-N
Synonym 5-bromo-3-iodobenzoic acid, 3-bromo-5-iodo-benzoic acid, 3-bromo-5-iodobenzoicacid, benzoic acid, 3-bromo-5-iodo, 3-iodo-5-bromobenzoic acid, pubchem3788, acmc-209erv, 3-bromo-5-iodobenzoic aicd, 3-bromo-5-iodo benzoic acid, 5-iodo-3-bromo-benzoic acid
PubChem CID 5182520
IUPAC Name 3-bromo-5-iodobenzoic acid
SMILES C1=C(C=C(C=C1Br)I)C(=O)O

Specifications

Melting Point 218°C to 221°C
Quantity 100 g
Beilstein 3240837
Sensitivity Light sensitive
Solubility Information Soluble in methanol. Sparingly Soluble (0.20g/L) (25°C),Calc.
Formula Weight 326.92
Percent Purity 97%
Chemical Name or Material 3-Bromo-5-iodobenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only