missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-5-fluorotoluene, 98%

Catalog No. p-7053834
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6154414 25 g
AAH6154422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6154414 Supplier Thermo Scientific Chemicals Supplier No. H6154414

CAS: 202865-83-6 | C7H6BrF | 189.027 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 202865-83-6
Molecular Formula C7H6BrF
Molecular Weight (g/mol) 189.027
MDL Number MFCD01861195
InChI Key CTNFNUQREIIROB-UHFFFAOYSA-N
Synonym 3-bromo-5-fluorotoluene, 3-fluoro-5-bromotoluene, 5-bromo-3-fluorotoluene, benzene, 1-bromo-3-fluoro-5-methyl, 1-bromo-3-fluoro-5-methyl-benzene, 3-bromo-5-fluoro toluene, pubchem3237, acmc-1cod9, ksc493k3l, tpc-i111
PubChem CID 2773396
IUPAC Name 1-bromo-3-fluoro-5-methylbenzene
SMILES CC1=CC(=CC(=C1)Br)F

Specifications

Color Colorless
Quantity 25 g
Formula Weight 189.02
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 3-Bromo-5-fluorotoluene
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only