missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-5-fluorophenol, 97%

Catalog No. p-7036854
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5528803 1 g
AAH5528806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5528803 Supplier Thermo Scientific Chemicals Supplier No. H5528803
Only null left

CAS: 433939-27-6 | C6H4BrFO | 190.999 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 433939-27-6
Molecular Formula C6H4BrFO
Molecular Weight (g/mol) 190.999
MDL Number MFCD07783710
InChI Key JCPJGUPQZDEZQH-UHFFFAOYSA-N
Synonym 3-fluoro-5-bromophenol, 5-bromo-3-fluorophenol, 3-bromo-5-fluoro-phenol, phenol, 3-bromo-5-fluoro, pubchem4129, acmc-1apwb, ksc493m6t, 3-bromo-5-fluorophenol
PubChem CID 21904636
IUPAC Name 3-bromo-5-fluorophenol
SMILES C1=C(C=C(C=C1F)Br)O

Specifications

Melting Point 36°C to 40°C
Flash Point >110°C (230°F)
Quantity 1 g
Formula Weight 191
Percent Purity 97%
Chemical Name or Material 3-Bromo-5-fluorophenol
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only