accessibility menu, dialog, popup

UserName

FBrOH

CAS RN 433939-27-6

IUPAC Name: 3-bromo-5-fluorophenol
Synonyms: 3-bromo-5-fluorophenol
Molecular Weight (g/mol): 191
Molecular Formula: C6H4BrFO
InChi Key: JCPJGUPQZDEZQH-UHFFFAOYSA-N
SMILES: OC1=CC(F)=CC(Br)=C1
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)