missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-5-chlorophenol, ≥97%, Thermo Scientific Chemicals

Catalog No. p-7056093
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6484406 5 g
AAH6484414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6484406 Supplier Thermo Scientific Chemicals Supplier No. H6484406

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 56962-04-0
Molecular Formula C6H4BrClO
Molecular Weight (g/mol) 207.45
MDL Number MFCD07780676
InChI Key GMGWXLPFRHYWAS-UHFFFAOYSA-N
Synonym phenol, 3-bromo-5-chloro, 3-bromo-5-chloro-phenol, 3-bromo-5-chlorophenol, 97+%, pubchem17565, acmc-209lv4, ksc268s7h
PubChem CID 22630180
IUPAC Name 3-bromo-5-chlorophenol
SMILES OC1=CC(Cl)=CC(Br)=C1

Specifications

Melting Point 66°C to 70°C
Quantity 5 g
Beilstein 3234780
Formula Weight 207.45
Percent Purity ≥97%
Physical Form Solid
Chemical Name or Material 3-Bromo-5-chlorophenol
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only