missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Bromo-5-chlorobenzonitrile 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 74°C |
| Color | White-Yellow |
| Boiling Point | 144°C |
| Quantity | 5 g |
| UN Number | 3439 |
| Formula Weight | 216.46 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 3-Bromo-5-chlorobenzonitrile |
Safety and Handling
| TSCA | No |