accessibility menu, dialog, popup

UserName

ClBrN

CAS RN 304854-55-5

IUPAC Name: 3-bromo-5-chlorobenzonitrile
Synonyms: 3-bromo-5-chlorobenzonitrile
Molecular Weight (g/mol): 216.46
Molecular Formula: C7H3BrClN
InChi Key: DRKWKPSNVQVDKZ-UHFFFAOYSA-N
SMILES: ClC1=CC(Br)=CC(=C1)C#N
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results