missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-5-chlorobenzoic Acid 98.0+%, TCI America™

Catalog No. p-7123400
Change view
Click to view available options
Quantity:
1 g
5 g
Catalog No. Quantity
B41911G 1 g
B41915G 5 g
2 options

Catalog No. B41911G

Supplier: TCI America B41911G

Only null left

Chemical Identifiers

CAS 42860-02-6
Molecular Formula C7H4BrClO2
Molecular Weight (g/mol) 235.46
MDL Number MFCD06797227
InChI Key PBRRUJWXRUGGAW-UHFFFAOYSA-N
PubChem CID 33128
IUPAC Name 3-bromo-5-chlorobenzoic acid
SMILES OC(=O)C1=CC(Cl)=CC(Br)=C1

Specifications

Melting Point 197°C
Color White-Yellow
Quantity 1 g
Formula Weight 235.46
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Bromo-5-chlorobenzoic Acid