accessibility menu, dialog, popup

UserName

CAS RN 42860-02-6

HOOClBr

CAS RN 42860-02-6

IUPAC Name: 3-bromo-5-chlorobenzoic acid
Synonyms: 3-bromo-5-chlorobenzoic acid
Molecular Weight (g/mol): 235.46
Molecular Formula: C7H4BrClO2
InChi Key: PBRRUJWXRUGGAW-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(Cl)=CC(Br)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results