missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-4-(trifluoromethoxy)aniline 97.0+%, TCI America™

Catalog No. b41905g
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B41905G 5 g
B41901G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B41905G Supplier TCI America Supplier No. B41905G

Chemical Identifiers

CAS 191602-54-7
Molecular Formula C7H5BrF3NO
Molecular Weight (g/mol) 256.022
MDL Number MFCD00190127
InChI Key RAQMUBDHNKQNTD-UHFFFAOYSA-N
Synonym 3-bromo-4-trifluoromethoxy aniline, 3-bromo-4-trifluoromethoxy benzenamine, 4-amino-2-bromo trifluoromethoxy benzene, 3-bromo-4-trifluoromethoxy-phenylamine, 2-bromo-4-amino-trifluoromethoxybenzene, benzenamine, 3-bromo-4-trifluoromethoxy, 1-amino-3-bromo-4-trifluoromethoxy benzene, pubchem2925, ksc496c0p
PubChem CID 2736418
IUPAC Name 3-bromo-4-(trifluoromethoxy)aniline
SMILES C1=CC(=C(C=C1N)Br)OC(F)(F)F

Specifications

Color Yellow
Quantity 5 g
Formula Weight 256.02
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3-Bromo-4-(trifluoromethoxy)aniline