accessibility menu, dialog, popup

UserName

NH2BrOFFF

CAS RN 191602-54-7

IUPAC Name: 3-bromo-4-(trifluoromethoxy)aniline
Synonyms: 3-bromo-4-(trifluoromethoxy)aniline
Molecular Weight (g/mol): 256.02
Molecular Formula: C7H5BrF3NO
InChi Key: RAQMUBDHNKQNTD-UHFFFAOYSA-N
SMILES: NC1=CC(Br)=C(OC(F)(F)F)C=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results