missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-4-methylbenzoic Acid 98.0+%, TCI America™

Catalog No. p-7119916
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B30495G 5 g
B304925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B30495G Supplier TCI America Supplier No. B30495G
Only null left

Chemical Identifiers

CAS 7697-26-9
Molecular Formula C8H7BrO2
Molecular Weight (g/mol) 215.05
MDL Number MFCD00002488
InChI Key ZFJOMUKPDWNRFI-UHFFFAOYSA-N
Synonym 3-bromo-4-methylbenzoicacid, 3-bromo-p-toluic acid, benzoic acid, 3-bromo-4-methyl, 3-bromo-4-methyl-benzoic acid, bromo-p-toluic acid, pubchem3806, bromo-para-toluic acid, 3-bromo4-methylbenzoic acid, acmc-1bg95, akos bbb/219
PubChem CID 82130
IUPAC Name 3-bromo-4-methylbenzoic acid
SMILES CC1=CC=C(C=C1Br)C(O)=O

Specifications

Melting Point 210°C
Quantity 5 g
Formula Weight 215.05
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Bromo-4-methylbenzoic Acid