accessibility menu, dialog, popup

UserName

CH3BrOHO

CAS RN 7697-26-9

IUPAC Name: 3-bromo-4-methylbenzoic acid
Synonyms: 3-bromo-4-methylbenzoic acid
Molecular Weight (g/mol): 215.05
Molecular Formula: C8H7BrO2
InChi Key: ZFJOMUKPDWNRFI-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1Br)C(O)=O
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)