missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Bromo-4-methylaniline 98.0+%, TCI America™
Supplier: TCI America B235425G
Specifications
| 3-Bromo-4-methylaniline | |
| 139°C | |
| MFCD00134176 | |
| 2811 | |
| GRXMMIBZRMKADT-UHFFFAOYSA-N | |
| 3-bromo-4-methylaniline | |
| 82187 | |
| ≥98.0% (GC,T) |
| 7745-91-7 | |
| C7H8BrN | |
| 25 g | |
| 3-bromo-p-toluidine, p-toluidine, 3-bromo, benzenamine, 3-bromo-4-methyl, 3-bromo-4-methyl-phenylamine, 3-bromo-1,4-toluidine, 4-amino-2-bromotoluene, 3-bromo-4-methylbenzenamine, 3-bromo-4-methylphenylamine, 3-bromo-para-toluidine, 3-bromo-4-methyl-aniline | |
| CC1=CC=C(N)C=C1Br | |
| 186.05 | |
| 186.05 | |
| Crystalline Lumps |
Chemical Identifiers
| 7745-91-7 | |
| 186.05 | |
| GRXMMIBZRMKADT-UHFFFAOYSA-N | |
| 82187 | |
| CC1=CC=C(N)C=C1Br |
| C7H8BrN | |
| MFCD00134176 | |
| 3-bromo-p-toluidine, p-toluidine, 3-bromo, benzenamine, 3-bromo-4-methyl, 3-bromo-4-methyl-phenylamine, 3-bromo-1,4-toluidine, 4-amino-2-bromotoluene, 3-bromo-4-methylbenzenamine, 3-bromo-4-methylphenylamine, 3-bromo-para-toluidine, 3-bromo-4-methyl-aniline | |
| 3-bromo-4-methylaniline |
Safety and Handling
RTECSNumber : XU4375000
TSCA : No