missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-4-methylaniline, 97%

Catalog No. p-7027699
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2467006 5 g
AAB2467014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2467006 Supplier Thermo Scientific Chemicals Supplier No. B2467006

CAS: 7745-91-7 | C7H8BrN | 186.05 g/mol

3-Bromo-4-methylaniline is used in the synthesis of 1,7-dihalo Tröger?s base isomers. It is suitable for use to investigate the reactions of distonic 4-(N,N,N-trimethylammonium)-2-methylphenyl and 5-(N,N,N-trimethylammonium)-2-methylphenyl radical cations with O2 by ion-trap mass spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Bromo-4-methylaniline is used in the synthesis of 1,7-dihalo Tröger′s base isomers. It is suitable for use to investigate the reactions of distonic 4-(N,N,N-trimethylammonium)-2-methylphenyl and 5-(N,N,N-trimethylammonium)-2-methylphenyl radical cations with O2 by ion-trap mass spectrometry.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 7745-91-7
Molecular Formula C7H8BrN
Molecular Weight (g/mol) 186.05
MDL Number MFCD00134176
InChI Key GRXMMIBZRMKADT-UHFFFAOYSA-N
Synonym 3-bromo-p-toluidine, p-toluidine, 3-bromo, benzenamine, 3-bromo-4-methyl, 3-bromo-4-methyl-phenylamine, 3-bromo-1,4-toluidine, 4-amino-2-bromotoluene, 3-bromo-4-methylbenzenamine, 3-bromo-4-methylphenylamine, 3-bromo-para-toluidine, 3-bromo-4-methyl-aniline
PubChem CID 82187
IUPAC Name 3-bromo-4-methylaniline
SMILES CC1=CC=C(N)C=C1Br

Specifications

Melting Point 25°C to 28°C
Boiling Point 254°C to 257°C
Flash Point >110°C (230°F)
Odor Characteristic
Refractive Index 1.6115
Quantity 5 g
UN Number UN2811
Beilstein 1562057
Solubility Information Insoluble in water.
Formula Weight 186.06
Percent Purity 97%
Chemical Name or Material 3-Bromo-4-methylaniline
Show More Show Less
Hazard Category H301-H315-H319-H335
Hazard Statement GHS H Statement
H301-H311-H315-H319
Toxic if swallowed.
Toxic in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 231-807-8
RTECSNumber XU4375000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only