missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-4-methoxytoluene 98.0+%, TCI America™

Catalog No. B30325G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B30325G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B30325G Supplier TCI America Supplier No. B30325G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 22002-45-5
Molecular Formula C8H9BrO
Molecular Weight (g/mol) 201.06
MDL Number MFCD00017783
InChI Key DHPUIKWBNXTXOB-UHFFFAOYSA-N
Synonym 2-Bromo-4-methylanisole
PubChem CID 89143
IUPAC Name 2-bromo-1-methoxy-4-methylbenzene
SMILES COC1=CC=C(C)C=C1Br

Specifications

Melting Point 16°C
Boiling Point 125°C
Quantity 5 g
Formula Weight 201.06
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Bromo-4-methoxytoluene