accessibility menu, dialog, popup

UserName

H3COCH3Br

CAS RN 22002-45-5

IUPAC Name: 2-bromo-1-methoxy-4-methylbenzene
Synonyms: 2-bromo-1-methoxy-4-methylbenzene
Molecular Weight (g/mol): 201.06
Molecular Formula: C8H9BrO
InChi Key: DHPUIKWBNXTXOB-UHFFFAOYSA-N
SMILES: COC1=CC=C(C)C=C1Br

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results