missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Bromo-4-fluorophenol 99.0+%, TCI America™
$78.29 - $311.30
Chemical Identifiers
| CAS | 27407-11-0 |
|---|---|
| Molecular Formula | C6H4BrFO |
| Molecular Weight (g/mol) | 190.999 |
| MDL Number | MFCD03425884 |
| InChI Key | QWTULQLVGNZMLF-UHFFFAOYSA-N |
| Synonym | 3-bromo-4-fluoro-phenol, phenol, 3-bromo-4-fluoro, pubchem1486, acmc-1cmha, ksc201s9b, 3-bromanyl-4-fluoranyl-phenol |
| PubChem CID | 2783381 |
| IUPAC Name | 3-bromo-4-fluorophenol |
| SMILES | C1=CC(=C(C=C1O)Br)F |
Chemical Identifiers
| 27407-11-0 | |
| 190.999 | |
| QWTULQLVGNZMLF-UHFFFAOYSA-N | |
| 2783381 | |
| C1=CC(=C(C=C1O)Br)F |
| C6H4BrFO | |
| MFCD03425884 | |
| 3-bromo-4-fluoro-phenol, phenol, 3-bromo-4-fluoro, pubchem1486, acmc-1cmha, ksc201s9b, 3-bromanyl-4-fluoranyl-phenol | |
| 3-bromo-4-fluorophenol |
Specifications
| 27407-11-0 | |
| 78°C | |
| MFCD03425884 | |
| 3-bromo-4-fluoro-phenol, phenol, 3-bromo-4-fluoro, pubchem1486, acmc-1cmha, ksc201s9b, 3-bromanyl-4-fluoranyl-phenol | |
| C1=CC(=C(C=C1O)Br)F | |
| 190.999 | |
| 191.00 | |
| Crystalline Powder |
| Yellow | |
| C6H4BrFO | |
| 5 g | |
| QWTULQLVGNZMLF-UHFFFAOYSA-N | |
| 3-bromo-4-fluorophenol | |
| 2783381 | |
| ≥99.0% (GC,T) | |
| 3-Bromo-4-fluorophenol |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator