accessibility menu, dialog, popup

UserName

CAS RN 27407-11-0

IUPAC Name: 3-bromo-4-fluorophenol
Synonyms: 3-bromo-4-fluorophenol
Molecular Weight (g/mol): 191
Molecular Formula: C6H4BrFO
InChi Key: QWTULQLVGNZMLF-UHFFFAOYSA-N
SMILES: OC1=CC(Br)=C(F)C=C1
Percent Purity 
  • (2)
  • (1)
Physical Form 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)