missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-4-fluorophenol 99.0+%, TCI America™

Catalog No. B44915G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B44915G 5 g
B449125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B44915G Supplier TCI America Supplier No. B44915G

Chemical Identifiers

CAS 27407-11-0
Molecular Formula C6H4BrFO
Molecular Weight (g/mol) 190.999
MDL Number MFCD03425884
InChI Key QWTULQLVGNZMLF-UHFFFAOYSA-N
Synonym 3-bromo-4-fluoro-phenol, phenol, 3-bromo-4-fluoro, pubchem1486, acmc-1cmha, ksc201s9b, 3-bromanyl-4-fluoranyl-phenol
PubChem CID 2783381
IUPAC Name 3-bromo-4-fluorophenol
SMILES C1=CC(=C(C=C1O)Br)F

Specifications

Color Yellow
Boiling Point 78°C
Quantity 5 g
Formula Weight 191.00
Percent Purity ≥99.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Bromo-4-fluorophenol
TSCA No
Recommended Storage Refrigerator